missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPNPEP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25945-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Aminopeptidase P is a hydrolase specific for N-terminal imido bonds, which are common to several collagen degradation products, neuropeptides, vasoactive peptides, and cytokines. Structurally, the enzyme is a member of the ′pita bread fold′ family and occurs in mammalian tissues in both soluble and GPI-anchored membrane-bound forms. A membrane-bound and soluble form of this enzyme have been identified as products of two separate genes.Especificaciones
XPNPEP2 | |
Polyclonal | |
Unconjugated | |
XPNPEP2 | |
mAmP, Membrane bound AmP, Membrane bound APP, X Pro aminopeptidase 2, Xaa Pro aminopeptidase 2, XPNPEP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7512 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43895 | |
XPNPEP2 | |
XPNPEP2 Fusion Protein Ag22952 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |