missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16462-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The XPA (xeroderma pigmentosumgroup A) protein specifically recognizes the UV-orchemically damaged DNA lesions, and triggers thenucleotide excision repair process. XPA binds to thereplication protein A (RPA) or the excision repaircross complementing 1 protein (ERCC 1). In the absence of nucleotide excision repair persisting (unrepaired) DNA lesions (adducts) may lead to the accumulation of gene mutations and ultimately to cancer. Xeroderma pigmentosum patients have a >2000 fold increased risk to develop skin cancer atsun-exposed areas.Especificaciones
XPA | |
Polyclonal | |
Unconjugated | |
XPA | |
XP1, XPA, XPAC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22590, 298074, 7507 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P23025, Q64267 | |
XPA | |
XPA Fusion Protein Ag9536 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |