missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSTM1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24382-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
VSTM1 (V-Set And Transmembrane Domain Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include cytokine activity. An important paralog of this gene is LILRB1.Especificaciones
VSTM1 | |
Polyclonal | |
Unconjugated | |
VSTM1 | |
UNQ3033, VSTM1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
284415 | |
-20°C | |
Liquid |
Western Blot, Flow Cytometry | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6UX27 | |
VSTM1 | |
VSTM1 Fusion Protein Ag15799 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |