missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSP15a Rabbit anti-Yeast, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 51090-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody recognizes the SPAPB1A10 protein of Yeast.
Especificaciones
VSP15a | |
Polyclonal | |
Unconjugated | |
SPAPB1A10.05 | |
SPAPB1A10.05 | |
VSP15a Fusion Protein Ag0724 | |
150 μL | |
Primary | |
Yeast | |
Antibody | |
IgG |
ELISA | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
SPAPB1A10, VSP15a | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2543600 | |
-20°C | |
Liquid |