Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette allow accurate measurements of a volume of solution.
Marca: Poulten & Graf GmbH R371/NC/F/5
Detalles adicionales : Peso : 0.17108kg
Descripción
- Amber graduation
- With 1 mark
- Conformity approved
- With batch certificate
Especificaciones
Bulb Pipette | |
A | |
Amber stain |
BS1583, ISO648 | |
10 mL |