Learn More
Vα2 TCR Rat anti-Mouse, FITC, Clone: B20.1, BD
Rat Monoclonal Antibody
Marca: BD Biosciences 562085
Detalles adicionales : Peso : 0.01000kg
Descripción
The B20.1 monoclonal antibody specifically binds to most members of the Vα2 T-cell Receptor (TCR) subfamily in mice having the a, b, and c haplotypes of the Tcrb α haplotypes. Moreover, the frequencies of Vα2+ CD8+ and CD4+ T cells are influenced by H-2 haplotypes.
Especificaciones
Vα2 TCR | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Soluble αβ TCR from mouse cytotoxic T-cell clone KB5-C20 | |
25 μg | |
Immunology | |
Mouse | |
IgG2a λ |
Flow Cytometry | |
B20.1 | |
FITC | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.