Learn More
Abnova™ USP15 Recombinant Protein
Human full-length ORF recombinant protein with GST-tag at N-terminal
Marca: Abnova™ H00009958-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Encoded by ubiquitin specific peptidase 15 gene
- Molecular weight: 51.59kDa
- Preparation method: in vitro wheat germ expression system
- Purification: glutathione sepharose 4 fast flow
- Storage buffer: 50 mM Tris-HCI, 10 mM reduced glutathione
- pH=8.0 in elution buffer
Sequence: MAEGGAADLDTQRSDIATLLKTSLRKGDTWYLVDSRWFKQWKKYVGFDSWDKYQMGDQNVYPGPIDNSGLLKDGDAQSLKEHLIDELDYILLPTEGWNKLVSWYTLMEGQEPIARKVVEQGMFVKHCKVEVYLTELKLCENGNMNNVVTRRFSKADTIDTIEKEIRKIFSIPDEKETRLWNKYMSNTF
EPLNKPDSTIQDAGLYQGQVLVIEQKNEDGTWPRGPSTPKKPLEQSC
Best when used within three months from the date of receipt of this protein.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Especificaciones
AAH20688 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
52 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MAEGGAADLDTQRSDIATLLKTSLRKGDTWYLVDSRWFKQWKKYVGFDSWDKYQMGDQNVYPGPIDNSGLLKDGDAQSLKEHLIDELDYILLPTEGWNKLVSWYTLMEGQEPIARKVVEQGMFVKHCKVEVYLTELKLCENGNMNNVVTRRFSKADTIDTIEKEIRKIFSIPDEKETRLWNKYMSNTFEPLNKPDSTIQDAGLYQGQVLVIEQKNEDGTWPRGPSTPKKPLEQSC | |
KIAA0529/MGC131982/MGC149838/MGC74854/UNPH4 | |
USP15 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
9958 | |
USP15 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
USP15 | |
Human | |
Recombinant | |
Solution |