missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Universal pH Indicator Paper
Proporciona lecturas rápidas y precisas. El papel indicador de pH universal Fisherbrand™ incluye una escala de color impresa en la tapa para mayor comodidad.
Marque: Fisherbrand™
Informations supplémentaires : Peso : 0.01000kg
Description
- Filter paper soaked with one or several colored indicators of pH
- Economic range of pH 0 to 14
- Available in a variety of formats to meet individual needs
- Reference chart shows color changes in steps of pH2
Spécification
De 0 a 10 | |
10 Pack | |
50 mm |
Tube | |
Grade 911 | |
10 mm |