missing translation for 'onlineSavingsMsg'
Learn More

UNC5C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 20240-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16809764

  • 399.00€ / 150 microlitros
Fecha estimada de envío: 22-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The antibody is specific to UNC5C.

This gene product belongs to the UNC-5 family of netrin receptors. Netrins are secreted proteins that direct axon extension and cell migration during neural development. They are bifunctional proteins that act as attractants for some cell types and as repellents for others, and these opposite actions are thought to be mediated by two classes of receptors. The UNC-5 family of receptors mediate the repellent response to netrin; they are transmembrane proteins containing 2 immunoglobulin -like domains and 2 type I thrombospondin motifs in the extracellular region.
Especificaciones

Especificaciones

UNC5C
Polyclonal
Unconjugated
UNC5C
Netrin receptor UNC5C, Protein unc 5 homolog 3, Protein unc 5 homolog C, unc 5 homolog C (C. elegans), UNC5C, UNC5H3
Rabbit
Antigen Affinity Chromatography
RUO
22253, 362049, 8633
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08747, O95185, Q761X5
Unc5c
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales