missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ UL32 (CMV) (P08318) Partial Recombinant phosphoProtein with GST-tag
Used for SDS-PAGE
Marca: Abnova™ P5154.500ug
Detalles adicionales : Peso : 0.00010kg
Especificaciones
P08318 | |
Lyophilized | |
UL32 (CMV) Recombinant Protein | |
Ion exchange column and HPLC reverse phase column | |
Store at -20°C on dry atmosphere. After reconstitution with distilled water, store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
UL32 | |
Recombinant | |
Escherichia coli expression system | |
>90% by SDS-PAGE |
SDS-PAGE | |
52kDa | |
Escherichia coli expression system | |
500 ug | |
RUO | |
E. coli | |
GST | |
Lyophilized |