missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBN2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18844-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Description
Ubinuclein 1 (UBN1) is a ubiquitously expressed evolutionarily conserved protein which binds to proliferation-promoting genes that are repressed by formation of senescence-associated heterochromatin foci (SAHF). Ubinuclein 1 associates with various transcription factors and with histone methyltransferase activity, is indispensable for SAHF formation and appears to be a regulator of senescence. Although in most cells ubinuclein is localized to the nucleus, in cells forming tight junctions it is recruited to the cell adhesion complexes, dependently on the cell density.Specifications
UBN2 | |
Polyclonal | |
Unconjugated | |
UBN2 | |
KIAA2030, ubinuclein 2, UBN2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
254048 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6ZU65 | |
UBN2 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |