missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2G2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10722-1-AP-20UL
Additional Details : Peso : 0.00010kg
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE2G2 is a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein shares 100% sequence identity with the mouse counterpart. This gene is ubiquitously expressed, with high expression seen in adult muscle.Specifications
UBE2G2 | |
Polyclonal | |
Unconjugated | |
UBE2G2 | |
UBC7, UBE2G2, Ubiquitin carrier protein G2, Ubiquitin protein ligase G2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22213, 294331, 7327 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P60604, P60605 | |
UBE2G2 | |
UBE2G2 Fusion Protein Ag1082 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |