missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15912-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
UBE1 catalyzes the first step in ubiquitin conjugation to mark cellular proteins for degradation. This gene complements an X-linked mouse temperature-sensitive defect in DNA synthesis, and thus may function in DNA repair. It is part of a gene cluster on chromosome Xp11.23. Alternative splicing results in 2 transcript variants encoding the same protein, but with different 5′ UTR.Especificaciones
UBE1 | |
Polyclonal | |
Unconjugated | |
UBA1 | |
A1S9, A1S9T, A1ST, AMCX1, GXP1, Protein A1S9, SMAX2, UBA1, UBA1A, UBE1, UBE1X, Ubiquitin activating enzyme E1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22201, 314432, 7317 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P22314, Q02053, Q5U300 | |
Uba1 | |
UBE1 Fusion Protein Ag8703 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |