Learn More
Transglutaminase II Ab-4, Rabbit Polyclonal Antibody,Lab Vision™ Epredia™
Provide accurate, reproducible results with the Epredia™ Transglutaminase II Ab-4, Rabbit Polyclonal Antibody.
Marca: Epredia RB-060-P0
Detalles adicionales : Peso : 1.00000kg
Descripción
TGase II is implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: Not determined
Immunogen: Recombinant full-length human TGase II
Molecular Weight: 77-85kDa
Positive Control: HUVEC cells, Breast CA
Cellular Localization: Cytoplasm, membrane and extracellular matrix Ab-4 is excellent for staining of endothelial cells in murine tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native verified)
Note:
USA: RUO; Int'l: RUO
Especificaciones
Transglutaminase II Ab-4 | |
Polyclonal | |
Unconjugated | |
Recombinant full length human TGase II | |
RUO | |
Primary | |
Concentrated |
Immunoprecipitation, Western Blot | |
1mg/mL | |
Rabbit | |
100 μL | |
Neuroscience | |
Human, Mouse | |
IgG |