missing translation for 'onlineSavingsMsg'
Learn More

trans-4-n-pentil-4'-(4-n-pentilciclohexil)bifenilo, 98 %, Thermo Scientific™

Catalog Number 15442716
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
134149-96-5
C28H40
376.63
MFCD22581225
GVZFNAQZYYUPER-UHFFFAOYSA-N
trans-4-n-pentyl-4'-4-n-pentylcyclohexyl biphenyl, 4-pentyl-4'-1s,4r-4-pentylcyclohexyl-1,1'-biphenyl
15606766
1-pentil-4-[4-(4-pentilciclohexil)fenil]benceno
CCCCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC=C(CCCCC)C=C1
Este artículo no se puede devolver. Vea la política de devoluciones
134149-96-5
C28H40
376.63
MFCD22581225
GVZFNAQZYYUPER-UHFFFAOYSA-N
trans-4-n-pentyl-4'-4-n-pentylcyclohexyl biphenyl, 4-pentyl-4'-1s,4r-4-pentylcyclohexyl-1,1'-biphenyl
15606766
1-pentil-4-[4-(4-pentilciclohexil)fenil]benceno
CCCCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC=C(CCCCC)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material trans-4-n-Pentyl-4'-(4-n-pentylcyclohexyl)biphenyl
Cantidad 1 g
Formula Weight (peso de la fórmula) 376.61
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'