missing translation for 'onlineSavingsMsg'
Learn More

Topoisomerase II α Ab-1, Rabbit Polyclonal Antibody,Lab Vision™ Epredia™

Provide accurate, reproducible results with the Epredia™ Topoisomerase II α Ab-1, Rabbit Polyclonal Antibody.

Marca:  Epredia RB-117-P1

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12692807

  • 492.00€ / 500 microlitros
Fecha estimada de envío: 21-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Topo IIα plays important roles in synthesis and transcription of DNA as well as chromosomal segregation during mitosis. It is reported to be a sensitive and specific marker of late S-, G2- & M-phases in transformed and developmentally regulated normal cells. Note that Ki67/M1B1 is also present in G1-phase. Topo IIα is also implicated in drug resistance of tumor cells.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: aa 1513-1530

Immunogen: A synthetic peptide, corresponding to aa 1513-1530 (RAKKPIKYLEESDEDDLF) from the C-terminus of human topoisomerase IIα

Molecular Weight: 170kDa

Positive Control: Raji cells or any exponentially growing cultured human cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Topoisomerase II alpha Ab-1
Polyclonal
Unconjugated
A synthetic peptide, corresponding to aa 1513-1530 (RAKKPIKYLEESDEDDLF) from the C-terminus of human topoisomerase IIalpha
RUO
Primary
Concentrated
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1mg/mL
Rabbit
500 μL
DNA Repair
Human
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales