missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOP2A Mouse anti-Human, Clone: 1E12B3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66541-1-IG-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Topo II alpha plays important roles in synthesis and transcription of DNA as well as chromosomal segregation during mitosis. It is reported to be a sensitive and specific marker of late S-, G2 and M-phases in transformed and developmentally regulated normal cellsTopo II alpha is also implicated in drug resistance of tumor cells.Especificaciones
TOP2A | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P11388 | |
TOP2A | |
TOP2A Fusion Protein Ag19551 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
1E12B3 | |
Unconjugated | |
TOP2A | |
DNA topoisomerase 2 alpha, TOP2, TOP2A, TP2A | |
Mouse | |
Protein A | |
RUO | |
7153 | |
-20°C | |
Liquid |