missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM9B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24331-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
TMEM9B, also termed as Transmembrane protein 9B encodes a 198 amino-acid protein that contains an N-terminal signal peptide, a single transmembrane region, three potential N-glycosylation sites, and three conserved cys-rich domains in the N-terminus. There is a dimer form of TMEM9B protein. TMEM9B mRNA is expressed in a wide range of tissues and cells. TMEM9B is a lysosomal transmembrane protein that regulates the activity of inflammatory signaling pathways.Especificaciones
TMEM9B | |
Polyclonal | |
Unconjugated | |
TMEM9B | |
C11orf15, TMEM9 domain family, member B, TMEM9B, Transmembrane protein 9B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
293415, 56674, 56786 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9JJR8, Q9NQ34 | |
TMEM9B | |
TMEM9B Fusion Protein Ag19383 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |