missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM27 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19207-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
TMEM27 (Transmembrane protein 27) is also named as collectrin and Belongs to the TMEM27 family. It is a stimulator of beta cell replication and a membrane protein cleaved and shed by pancreatic beta cells that has been proposed as a beta cell mass biomarker. Western blot analysis of mouse collecting duct cell lines detected Tmem27 at an apparent molecular mass of 32 kDa. TMEM27 is an N-glycosylated protein and forms dimers, with MW of 64-75 kDa.Especificaciones
TMEM27 | |
Polyclonal | |
Unconjugated | |
TMEM27 | |
ACE2, Collectrin, NX 17, NX17, TMEM27, transmembrane protein 27 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57393, 57394 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9ESG4, Q9HBJ8 | |
TMEM27 | |
TMEM27 Fusion Protein Ag7668 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |