Learn More
TIPIN Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11972-2-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The function of this protein remains unknown.Especificaciones
TIPIN | |
Polyclonal | |
Unconjugated | |
TIPIN | |
TIMELESS interacting protein, TIPIN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54962 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BVW5 | |
TIPIN | |
TIPIN Fusion Protein Ag2595 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |