missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26647-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintece of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.Especificaciones
TIMP2 | |
Polyclonal | |
Unconjugated | |
TIMP2 | |
CSC 21K, Metalloproteinase inhibitor 2, TIMP 2, TIMP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21858, 7077 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P16035, P25785 | |
TIMP2 | |
TIMP2 Fusion Protein Ag24436 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |