Learn More
Thermo Scientific™ Thermal Printer and Paper for Eutech™ Meters
Thermal Printer for Eutech™ meters provides the convenience of printing directly from your meter whenever, wherever.
Marca: Thermo Scientific™ 01X594401
Detalles adicionales : Peso : 0.20000kg
Descripción
Thermal Printer for Eutech™ meters provides the convenience of printing directly from your meter whenever, wherever.This compact thermal printer connects easily to your 110 series handheld, 450 series waterproof handheld or 2700 series benchtop meters via RS232 – ideal for data-recording and report-generation. Connecting cables included.
Características:
- Térmica
- Incluye rollo de papel
- Modelo con reloj en tiempo real y adaptador RS232 disponible
Especificaciones
Papel de recambio para impresora térmica, dos rollos | |
Pieza de repuesto |