missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TESPA1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25975-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
TESPA1 is a critical component of the TCR signalosome and is essential for T cell selection and maturation through the regulation of TCR signaling during T cell development. TESPA1 encodes a protein of 458 amino acids and is highly conserved among vertebrate species. TESPA1 expression was tightly regulated during T cell development, with highest expression at the DP stage. TESPA1 deficiency resulted in failure of positive selection and substantially fewer of CD4+ and CD8+ SP cells in the thymus.Especificaciones
TESPA1 | |
Polyclonal | |
Unconjugated | |
TESPA1 | |
HSPC257, KIAA0748, TESPA1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9840 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A2RU30 | |
TESPA1 | |
TESPA1 Fusion Protein Ag23218 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |