missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TC10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17805-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
TC10 is a member of the RAS superfamily of small GTP-binding proteins (see HRAS, MIM 190020) involved in insulin-stimulated glucose uptake.Especificaciones
TC10 | |
Polyclonal | |
Unconjugated | |
RHOQ | |
ARHQ, Ras like protein TC10, RASL7A, RHOQ, TC10, TC10A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23433 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P17081 | |
RHOQ | |
TC10 Fusion Protein Ag12078 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |