missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBXA2R Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27159-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The thromboxane A2 receptor (TBXA2R) is a member of the Prostanoid Receptor subfamily. TBXA2R performs an essential role in hemostasis by interacting with thromboxane A2 to induce platelet aggregation. Thromboxane A2 has been implicated in myocardial infarct, stroke, and asthma. Additionally, thromboxane-prostanoid receptors have been implicated in the pathogenesis of cardiovascular diseases. Changes in TBXA2R function have also been linked to a hereditary bleeding disorder.Especificaciones
TBXA2R | |
Polyclonal | |
Unconjugated | |
TBXA2R | |
Prostanoid TP receptor, TBXA2R, thromboxane A2 receptor, TXA2 R | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21390, 6915 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P21731, P30987 | |
TBXA2R | |
TBXA2R Fusion Protein Ag25670 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |