missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBL1XR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 55312-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene has sequence similarity with members of the WD40 repeat-containing protein family. The WD40 group is a large family of proteins, which appear to have a regulatory function. It is believed that the WD40 repeats mediate protein-protein interactions and members of the family are involved in signal transduction, RNA processing, gene regulation, vesicular trafficking, cytoskeletal assembly and may play a role in the control of cytotypic differentiation.Especificaciones
TBL1XR1 | |
Polyclonal | |
Unconjugated | |
TBL1XR1 | |
C21, DC42, IRA1, TBL1 related protein 1, TBL1XR1, TBLR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79718, 81004 | |
-20°C | |
Liquid |
Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BHJ5, Q9BZK7 | |
TBL1XR1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |