missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF6L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15745-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
In eukaryotic systems, the process of initiating transcription from protein-coding genes requires the presence of RNA polymerase II and a broad family of auxiliary transcription factors. TFIID is a general transcription factor that initiates preinitiation complex assembly through direct interaction with the TATA promoter element. Functioning as a multisubunit complex consisting of a small TATA-binding polypeptide and other TBP-associated factors (TAFs), TFIID mediates promoter responses to various transcriptional activators and repressors. TAF6L, also known as PAF65A, is a 622 amino acid nuclear protein that functions as part of the PCAF (p300/CBP-associated factor) histone acetylase complex.Especificaciones
TAF6L | |
Polyclonal | |
Unconjugated | |
TAF6L | |
PAF65 alpha, PAF65A, TAF6L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10629, 225895, 309194 | |
-20°C | |
Liquid |
Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8R2K4, Q9Y6J9 | |
TAF6L | |
TAF6L Fusion Protein Ag8180 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |