missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SURF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16522-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene shares a bidirectional promoter with surfeit 1 (SURF1; GeneID: 6834), which is located on the opposite strand. It encodes a conserved protein that is expressed in a variety of tissues.Especificaciones
SURF2 | |
Polyclonal | |
Unconjugated | |
SURF2 | |
SURF 2, SURF2, surfeit 2, Surfeit locus protein 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20931, 619345, 6835 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P09926, Q15527 | |
Surf2 | |
SURF2 Fusion Protein Ag9745 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |