missing translation for 'onlineSavingsMsg'
Learn More

SULT1A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10911-2-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16827613

  • 140.00€ / 20 microlitros
Fecha estimada de envío: 21-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes one of two phenol sulfotransferases with thermostable enzyme activity. Multiple alternatively spliced variants that encode two isoforms have been identified for this gene.
Especificaciones

Especificaciones

SULT1A1
Polyclonal
Unconjugated
SULT1A1
Aryl sulfotransferase 1, HAST1/HAST2, P PST, P PST 1, Phenol sulfotransferase 1, PST, ST1A1, ST1A3, STP, STP1, Sulfotransferase 1A1, SULT1A1, Ts PST, TSPST1
Rabbit
Antigen Affinity Chromatography
RUO
20887, 6817, 83783
-20°C
Liquid
Immunoprecipitation, Flow Cytometry, Western Blot, Immunocytochemistry, Immunofluorescence
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P17988, P50225, P52840
SULT1A1
SULT1A1 Fusion Protein Ag1344
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales