missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUCLG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14923-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes the alpha subunit of the heterodimeric enzyme succinate coenzyme A ligase. This enzyme is targeted to the mitochondria and catalyzes the conversion of succinyl CoA and ADP or GDP to succinate and ATP or GTP. Mutations in this gene are the cause of the metabolic disorder fatal infantile lactic acidosis and mitochondrial DNA depletion.Especificaciones
SUCLG1 | |
Polyclonal | |
Unconjugated | |
SUCLG1 | |
G ALPHA, SCS alpha, SUCLA1, SUCLG1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114597, 56451, 8802 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P13086, P53597, Q9WUM5 | |
Suclg1 | |
SUCLG1 Fusion Protein Ag6481 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |