missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRPK2 Recombinant Human Protein
SRPK2 Recombinant Human Protein
Marca: Invitrogen PV3829
Detalles adicionales : Peso : 0.05250kg
Descripción
SRPK2, along with SRPK1, is most likely the cellular protein kinase mediating HBV core protein phosphorylation during viral infection. It therefore represents an important host cell target for therapeutic intervention in HBV infection.Especificaciones
NP_872633 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
105kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
SRPK2, SFRSK2 | |
SRPK2 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 100μM RS domain derived peptide substrate (GRSRSRSRSR) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
SRPK2 Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.29mg/mL | |
3569, 6733 | |
SRPK2 Recombinant Human Protein | |
–80°C | |
1 Each | |
SRPK2 Recombinant Human Protein | |
SRPK | |
2,380nmoles of phosphate transferred to RS domain derived peptide substrate (GRSRSRSRSR) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.83μg/mL | |
GST Tag | |
Kinase and inhibitors | |
70% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.