missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24980-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
SPATA1 is a protein coding gene.Especificaciones
SPATA1 | |
Polyclonal | |
Unconjugated | |
SPATA1 | |
SP 2, SPAP1, SPATA1, Sperm specific protein SP 2, spermatogenesis associated 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100505741 | |
-20°C | |
Liquid |
Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5VX52 | |
SPATA1 | |
SPATA1 Fusion Protein Ag21784 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |