Learn More
Gel de sílice y sodio, 35 a 40 % de metal alcalino en gel de sílice, fase I, Thermo Scientific™
Produce aniones y dianiones radicales por reducción, desclora haluros de alquilo y arilo y desulfura compuestos
Marca: Thermo Scientific Alfa Aesar 044893.22
Detalles adicionales : Peso : 0.10000kg Transport : UN number : 2813 Chem class : 4.3 Pack group : II
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Especificaciones
Sodium-silica gel | |
Negro | |
100 g | |
Reacts with water. | |
0.7 |
35-40% alkali metal in silica gel, Stage I | |
Polvo | |
MFCD01941718 | |
Moisture sensitive |