missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A12 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26804-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a calcium-binding mitochondrial carrier protein. The encoded protein localizes to the mitochondria and is involved in the exchange of aspartate for glutamate across the inner mitochondrial membrane. Polymorphisms in the gene encoding this protein may be associated with risk of autism.Especificaciones
SLC25A12 | |
Polyclonal | |
Unconjugated | |
SLC25A12 | |
AGC1, ARALAR, ARALAR1, SLC25A12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
78830, 8604 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75746, Q8BH59 | |
SLC25A12 | |
SLC25A12 Fusion Protein Ag25150 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |