missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-H Rat anti-Mouse, BUV563, Clone: 440c, BD Biosciences™
Rat Monoclonal Antibody
Marca: BD Biosciences 748295
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Siglec-H Monoclonal antibody specifically detects Siglec-H in Mouse samples. It is validated for Flow Cytometry
Especificaciones
| Siglec-H | |
| Monoclonal | |
| Brilliant Ultraviolet 563 | |
| Siglec H; Siglech; Sialic acid binding Ig-like lectin H; SIGLEC | |
| Mouse Bone Marrow IPC | |
| RUO | |
| Mouse | |
| IgG2b κ |
| Flow Cytometry | |
| 440c | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV563 under optimal conditions that minimize unconjugated dye and antibody. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Siglec-H Rat anti-Mouse, BUV563, Clone: 440c, BD Biosciences™ > 50 μg; BUV563
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido

