missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SGKL (SGK3), GST Tag Recombinant Protein
Human SGKL (SGK3), GST Tag Recombinant Protein
Marca: Invitrogen™ PV4058
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Unlike SGK, SGKL is not induced by serum or glucocorticoids. It is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK.
Especificaciones
NP_037389, S487D | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
SGKL (SGK3) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
74.3kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
SGK Kinase Family | |
Recombinant | |
Baculovirus |