missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SFN (Human) Recombinant Protein
Human SFN partial ORF ( AAH00329.1, 3 a.a. - 238 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00002810-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: RASLIQKAKLAEQAERYEDMAAFMKGAVEKGEELSCEERNLLSVAYKNVVGGQRAAWRVLSSIEQKSNEEGSEEKGPEVREYREKVETELQGVCDTVLGLLDSHLIKEAGDAESRVFYLKMKGDYYRYLAEVATGDDKKRIIDSARSAYQEAMDISKKEMPPTNPIRLGLALNFSVFHYEIANSPEEAISLAKTTFDEAMADLHTLSEDSYKDSTLIMQLLRDNLTLWTADNAGEE
Especificaciones
AAH00329.1 | |
stratifin | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SFN | |
GST |
2810 | |
Wheat germ expression system | |
25 μg | |
YWHAS | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |