missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SEPT5 (Human) Recombinant Protein
Human 40791 full-length ORF ( AAH25261, 1 a.a. - 369 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00005413-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MSTGLRYKSKLATPEDKQDIDKQYVGFATLPNQVHRKSVKKGFDFTLMVAGESGLGKSTLVHSLFLTDLYKDRKLLSAEERISQTVEILKHTVDIEEKGVKLKLTIVDTPGFGDAVNNTECWKPITDYVDQQFEQYFRDESGLNRKNIQDNRVHCCLYFISPFGHGLRPVDVGFMKALHEKVNIVPLIAKADCLVPSEIRKLKERIREEIDKFGIHVYQFPECDSDEDEDFKQQDRELKESAPFAVIGSNTVVEAKGQRVRGRLYPWGIVEVENQAHCDFVKLRNMLIRTHMHDLKDVTCDVHYENYRAHCIQQMTSKLTQDSRMESPIPILPLPTPDAETEKLIRMKDEELRRMQEMLQRMKQQMQDQ
Especificaciones
AAH25261 | |
septin 5 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SEPT5 | |
GST |
5413 | |
Wheat germ expression system | |
25 μg | |
CDCREL, CDCREL-1, CDCREL1, H5, PNUTL1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |