Learn More
ACROBiosystems SARS-CoV-2 S protein RBD (N501Y), Fc Tag
Marca:  ACROBiosystems SPD-C5253-1mg
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 17216510
  • 3495.90€ / 1 miligramo
Fecha estimada de envío: 15-05-2024
para ver el stock
Descripción
Descripción
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.1 kDa.
Especificaciones
Especificaciones
51.1 kDa
SARS-CoV-2 S protein RBD (N501Y), Fc Tag (SPD-C5253) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (N501Y)).
-20°C
S protein RBD
SARS-CoV-2
HEK293
Viral Proteins
1 mg
Arg 319 - Lys 537
Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19
Unconjugated
Fc Tag
Powder