Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S1 protein CTD, His Tag
MALS verified
Marca: ACROBiosystems S1D-C52H3-1mg
Detalles adicionales : Peso : 0.00010kg
Descripción
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.6 kDa. The protein migrates as 28-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
QHD43416.1 | |
1 mg | |
Asn 334 - Pro 527 | |
S1 protein CTD,Spike protein S1 CTD,BetaCoV S1-CTD,COVID-19 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
23.6 kDa | |
SARS-CoV-2 S1 protein CTD, His Tag (MALS-verified) (S1D-C52H3) is expressed from human 293 cells (HEK293). It contains AA Asn 334 - Pro 527 (Accession # QHD43416.1). | |
-20°C | |
S1 protein CTD | |
SARS-CoV-2 | |
PBS,pH7.3 | |
HEK293 | |
Viral Proteins |