missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RTN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11168-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene belongs to the family of reticulon encoding genes. Reticulons are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified.Especificaciones
RTN2 | |
Polyclonal | |
Unconjugated | |
RTN2 | |
NSP like protein 1, NSP like protein I, NSP2, NSPL1, NSPLI, reticulon 2, RTN2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20167, 308410, 6253 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O70622, O75298 | |
RTN2 | |
RTN2 Fusion Protein Ag1655 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |