missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPP21 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16377-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Especificaciones
RPP21 | |
Polyclonal | |
Unconjugated | |
RPP21 | |
C6orf135, CAT60, Ribonucleoprotein V, RNaseP protein p21, RPP21 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
406230, 67676, 79897 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8R040, Q9H633 | |
Rpp21 | |
RPP21 Fusion Protein Ag9353 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |