missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RILPL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16732-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Neuroprotective protein, which acts by sequestring GAPDH in the cytosol and prevent the apoptotic function of GAPDH in the nucleus. Competes with SIAH1 for binding GAPDH. Does not regulate lysosomal morphology and distribution. Plays a role in the regulation of cell shape and polarity. Plays a role in cellular protein transport, including protein transport away from primary cilia.Especificaciones
RILPL1 | |
Polyclonal | |
Unconjugated | |
RILPL1 | |
RILP like protein 1, RILPL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
304469, 353116, 75695 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
D3ZUQ0, Q5EBL4, Q9JJC6 | |
RILPL1 | |
RILPL1 Fusion Protein Ag10192 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |