missing translation for 'onlineSavingsMsg'
Learn More

RHBDL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 12467-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16830194

  • 140.00€ / 20 microlitros
Fecha estimada de envío: 15-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

RHBDL2 is a member of the rhomboid family of integral membrane proteins. This family contains proteins that are related to Drosophila rhomboid protein. Members of this family are found in both prokaryotes and eukaryotes and are thought to function as intramembrane serine proteases. RHBDL2 is thought to release soluble growth factors by proteolytic cleavage of certain membrane-bound substrates, including ephrin B2 and ephrin B3.
Especificaciones

Especificaciones

RHBDL2
Polyclonal
Unconjugated
RHBDL2
RHBDL2, Rhomboid like protein 2, Rhomboid related protein 2, RRP2
Rabbit
Antigen Affinity Chromatography
RUO
230726, 298512, 54933
-20°C
Liquid
Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A2AGA4, Q9NX52
RHBDL2
RHBDL2 Fusion Protein Ag3142
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales