missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFTN1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24289-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The function remains unknown.The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s) and ubiquitin-protein ligases (E3s). This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is demonstrated to participate in the ubiquitination of p53, c-Fos, and the NF-kB precursor p105 in vitro. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Especificaciones
RFTN1 | |
Polyclonal | |
Unconjugated | |
RFTN1 | |
KIAA0084, MIG2, PIB10, PIG9, Raft linking protein, RAFTLIN, raftlin, lipid raft linker 1, RFTN1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23180 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14699 | |
RFTN1 | |
RFTN1 Fusion Protein Ag17264 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |