missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15813-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Riboflavin kinase is an essential enzyme that catalyzes the phosphorylation of riboflavin to form flavin mononucleotide, an obligatory step in vitamin B2 utilization and flavin cofactor synthesis.Especificaciones
RFK | |
Polyclonal | |
Unconjugated | |
RFK | |
Flavokinase, RFK, riboflavin kinase, RIFK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
499328, 54391, 55312 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CFV9, Q969G6 | |
Rfk | |
RFK Fusion Protein Ag8591 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |