missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Virus Parvovirus B19 VLP VP1 Protein
A cDNA sequence encoding the Parvovirus B19 VLP VP1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12966-5000ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Virus Parvovirus B19 VLP VP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Parvovirus B19 VLP VP1 protein solution is supplied in PBS pH 7.4 and 0.02% Sodium azide. |
5 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Virus | |
Untagged | |
Greater than 90.0% as determined by 12% PAGE (coomassie staining). |