missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Toxoplasma Gondii Toxoplasma P22 Protein
A cDNA sequence encoding the Toxoplasma P22 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13787-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Toxoplasma Gondii Toxoplasma P22 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Sterile Filtered solution containing Phosphate buffered saline. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Toxoplasma Gondii | |
His | |
Toxoplasma protein is >95% pure as determined by 12% PAGE (coomassie staining). |