Learn More
enQuireBio™ Recombinant SARS SARS Mosaic S(N) Protein
Recombinant Protein
Marca: enQuireBio™ QP13422-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SARS SARS Mosaic S(N) Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-HCl, 0.4% sarcosyl, 0.25% Triton .–100 and 50% glycerol. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
SARS | |
Untagged | |
SARS Mosaic Protein is >95% pure as determined by 10% PAGE (coomassie staining). |