missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae IFM1, mitochondrial Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) IFM1, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7172-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_014619.1 | |
S. cerevisiae IFM1, mitochondrial Protein | |
Research Use Only | |
IFM1 | |
Recombinant Protein | |
E. coli | |
KGKRRNQISK KELKPLNFSI PNYISVNKLA NLLNCRVERL IKDLTALGFE NITTTYILSK EYVELILQEY NFALPNLSTS TNLDNVYDEL KSPVNPKLLT KRAPVVTIMG HVDHGKTTII DYLRKSSVVA QEHGGITQHI GAFQITAPKS GKKITFLDTP GHAAFLKMRE RGANITDIIV LVVSVEDSLM PQTLEAIKHA KNSGNEMIIA ITKIDRIPQP KEREKKIEKV INDLIVQGIP VEKIGGDVQV IPISAKTGEN MDLLEESIVL LSEVMDIRAE NSPKTIAEGW IIESQVKKQV GNVATVLVKK GTLQKGKILI CGNTFCKIKN LIDDKGIPIL KATPSYATEV LGWKDVPHVG DEVIQVKSEA IAKKFISKRQ DLIEVQKNSS IVEKLNEERA LAKEQHLNKE LEHENTVQEH EQNTGPKLIN YIIKCDVSGS AEAVSESISS LGNDEVRCNV ISSSVGIPTE SDLKMAQITE STILCFNLGN LPSEVINNRA GIKIKQYNVI YKLIEDVTET LTENLKPIFE KKIVSTVDVR ETFDFRLKKK IIRIAGCKVN NGVIKKNSLV QVVRGPNEDV IFDGKISTLK HNKDDVAEVS KGHECGITFE SGFEGFKPGD KILVYENVRV PRYL | |
Greater than 90% as determined by SDS-PAGE. |
854135 | |
500 μg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
GST | |
Tris-based buffer, 50% glycerol |